Chemical resistant shoe covers, MICROGARD® 2500, model 400
Catalog # ANSEWH25B-00400-00
Anbieter: Ansell
Technische Daten
- Shoe size:42 - 46
- Garment application:Laboratory
- Colour:White
- Pk:350 PAIR
Technische Daten
Über diesen Artikel
These over shoes are made of microporous PP film with a non woven inner layer. The fabric provides an excellent barrier to chemical spray and infective agents.
- Elasticated opening
- Bound seams
- Fit shoe size 42 - 46