Zwitterionic columns, CHIRALPAK® ZWIX
The CHIRALPAK® ZWIX columns were developed for separation of a great variety of zwitterionic molecules, specifically, for separation of underivatised amino acids.
ZWIX selectors are zwitterionic molecules that incorporate both anion- and cation-exchange functional groups. These ZWIX selectors exhibit exceptional enantioseparation capabilities toward zwitterionic molecules such as amino acids and small peptides.
Frequently Bought Together