Anti-CA9 Mouse Monoclonal Antibody [clone: PN-15]
ABNOMAB14416
: Abnova
- Antibody type:Primary
- Antigen name:Microsomal fraction renal cortical tissue homogenate
- Antigen symbol:CA9
- Clonality:Monoclonal
- Clone:PN-15
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:768
- Storage buffer:In PBS (0.05% BSA, 0.05% sodium azide).
- Storage temperature:Store at 4° C.
- Immunogen:Microsomal fraction of human renal cortical tissue homogenate.
- Purification:Protein A purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against native human CA9.
Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: PN-15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human