Real-Time PCR kit for detection of Vibrio alginolyticus
Supplier: Q BIOANALYTIC
Certificates
About this item
Vibrio alginolyticus causes eye, ear and wound infections. It can be tested using this Real-Time PCR test. The analysis can be done in water and food samples. You can use this test in combination with an appropriate pre- enrichment in alkaline peptone-water or you can confirm bacterial colonies on agar plates.
- TaqMan probes
- Contains internal amplification control in HEX channel
- Taget in FAM channel
- Contains UNG to protect from re-amplification
- Positive DNA and negative control included
Certifications: DIN EN ISO 20838 conform detection in food, feed and water samples from pre-enrichment cultures or for confirmation of bacterial colonies.
Frequently Bought Together