ContactUsLinkComponent
HyClone™ Dialysed Foetal Bovine Serum, US origin
HyClone™ Dialysed Foetal Bovine Serum, US origin
Certificates
undefined
HyClone™ Dialysed Foetal Bovine Serum, US origin
Specifications
  • Catalog No:
  • SH30079.03IR25
  • HYCLSH30079.01
  • HYCLSH30079.03HI
  • HYCLSH30079.03
  • HYCLSH30079.02
  • HYCLSH30079.01HI
  • HYCLSH30079.02HI
  • Storage temperature:
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.