Order Entry
Switzerland
ContactUsLinkComponent
 
Upright microscopes, BA210 and BA310
 :  MOTIC
 
undefined
Upright microscopes, BA210 and BA310
 :  MOTIC

 

  • Catalog No:
  • 630-2828
  • 630-2739
  • MOTI1100100401487
  • MOTI1100100401515
  • Head:
  • Siedentopf, 30° inclined, 360° rotating
  • Siedentopf, 30° inclined, 360° rotating
  • Siedentopf, 30° inclined, 360° rotating
  • Siedentopf, 30° inclined, 360° rotating
  • Eyepieces:
  • WF 10×/20 mm with pointer adjustment
  • WF 10×/20 mm with pointer adjustment
  • WF 10×/20 mm with pointer adjustment
  • WF 10×/20 mm with pointer adjustment
  • Stage:
  • 140×135 mm (movement: 76×50 mm)
  • 140×135 mm (movement: 76×50 mm)
  • 140×135 mm (movement: 76×50 mm)
  • 140×135 mm (movement: 76×50 mm)
  • Condenser:
  • Abbe 1,25 N.A.
  • Abbe 1,25 N.A.
  • Abbe 1,25 N.A.
  • Abbe 1,25 N.A.
  • Focusing system:
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Illumination:
  • LED
  • LED
  • LED
  • LED

 

Eyepieces

 

Frequently Bought Together