Mini tube system, Axygen®
Supplier: Corning
Certificates
About this item
PP, 100% virgin. The Axygen 96-well mini tube system is designed to work with 8-strip or 12-strip plug caps.
- Compatible with Beckman, Tecan, Zymark and other robotic workstations
- One-way lid design with etched in alphnumeric grid ensures positive sample identification
- Moulded in condensation rings on cover minimise risk of cross-contamination
Frequently Bought Together