ContactUsLinkComponent
HPTLC Vario system
HPTLC Vario system
Catalog # 552-0049
Supplier:  CAMAG
undefined
HPTLC Vario system
Catalog # 552-0049
Supplier:  CAMAG
Specifications
  • Description:
    HPTLC Vario system
  • Pk:
    1
Specifications
About this item
Time saving by optimisation of development conditions.
  • Development with six different solvents can be tested side by side
  • Both sandwich and tank configurations can be simulated side by side, making results directly comparable
  • Six different conditions of pre-equilibration, including relative humidity, can be tested simultaneously
  • These variations of developing conditions can be freely combined
Delivery: The HPTLC Vario system comprises an HPTLC Vario chamber for 100×100 mm plates and an HPTLC scoring unit for preparation of TLC/HPTLC plates.