Order Entry
Switzerland
ContactUsLinkComponent
 
Anion exchange chromatography columns, TSK-GEL®, BioAssist Q
Catalog # TOYO0019685
Supplier:  TOSOH Bioscience
undefined
Anion exchange chromatography columns, TSK-GEL®, BioAssist Q
Catalog # TOYO0019685
Supplier:  TOSOH Bioscience

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    10 µm
  • Pore size:
    4000 Å
  • Description:
    Anion exchange chromatography column, BioAssit Q
  • Length:
    50 mm
  • Column hardware:
    PEEK
  • Phase:
    BioAssit Q
  • Pk:
    1

Specifications

About this item

TSK-Gel® BioAssist Q are polymer based packed PEEK columns for laboratory applications. These columns provide high capacity for small to very large MW proteins and nucleic acids.

  • High capacity even for large proteins
  • Unique pore structure