Cannon-Fenske viscometers for manual measurements
Supplier: SI Analytics
Certificates
About this item
Cannon-Fenske viscometers for manual measurements especially in routine analyses.
Suitable for transparent liquids, foaming liquids and liquid mixture with highly volatile components.
Frequently Bought Together