Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stirrers
Stirring Rods
Stirring rods
Stirring rods
Stirring rods
Catalog # HECH42067001
Supplier:
GLASWARENFABRIK KARL HECHT
Expand image
Stirring rods
Catalog # HECH42067001
Supplier:
GLASWARENFABRIK KARL HECHT
Specifications
Ø:
2,5 mm
Description:
Stirring rod, width 3 mm
Material:
Plastic
Pk:
500
Specifications
About this item
Product Details
Product Details