Order Entry
Switzerland
ContactUsLinkComponent
Streptomyces avidini Streptavidin, FITC (Fluorescein Isothiocyanate), MP Biomedicals
Streptomyces avidini Streptavidin, FITC (Fluorescein Isothiocyanate), MP Biomedicals
Catalog # ICNA08623411
Supplier:  MP Biomedicals
CAS Number:  
Streptomyces avidini Streptavidin, FITC (Fluorescein Isothiocyanate), MP Biomedicals
Catalog # ICNA08623411
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Species:
    Streptomyces avidinii
  • Storage conditions:
    0 to +5 °C
  • Protein/peptide name:
    Streptavidin
  • Formulation:
    Liquid

Specifications

About this item

Streptavidin-FITC conjugated can be used for the detection or localisation of biotinylated biomolecules. It is purified by molecular sieving and is completely free of unbound FITC.

  • Working dilution: 1:50 to 1:150