Biotin-NHS
Catalog # 203118-50
Supplier: Merck Millipore (Oncogene)
CAS Number:
Specifications
- Pk:50 mg
- Description:Biotin-NHS, water-soluble
Specifications
About this item
Biotin-NHS reacts with primary amines to biotinylate proteins under mild conditions. Since it is water-soluble, this compound can be used to biotinylate cell-surface proteins and proteins that are sensitive to organic solvents.