ContactUsLinkComponent
Freund's incomplete adjuvant, MP Biomedicals
Freund's incomplete adjuvant, MP Biomedicals
Catalog # ICNA08642861
Supplier:  MP Biomedicals
CAS Number:  
Freund's incomplete adjuvant, MP Biomedicals
Catalog # ICNA08642861
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    50 mL
  • Description:
    Freund's incomplete adjuvant
Specifications
About this item
Freund's incomplete adjuvant mixture contains mineral oil and Arlacel A.
  • Enhances immune response in booster injections
  • Water-in-oil emulsion without Mycobacterium