Order Entry
Switzerland
ContactUsLinkComponent
 
Chest wader, Glenroe 700A
 :  SIOEN
 
undefined
Chest wader, Glenroe 700A
 :  SIOEN

 

  • Catalog No:
  • SIOE700AA2B17A9639
  • SIOE700AA2B17A9647
  • SIOE700AA2B17A9646
  • SIOE700AA2B17A9641
  • SIOE700AA2B17A9640
  • SIOE700AA2B17A9642
  • SIOE700AA2B17A9644
  • SIOE700AA2B17A9643
  • SIOE700AA2B17A9645
  • Model:
  • Glenroe 700A
  • Glenroe 700A
  • Glenroe 700A
  • Glenroe 700A
  • Glenroe 700A
  • Glenroe 700A
  • Glenroe 700A
  • Glenroe 700A
  • Glenroe 700A
  • Garment application:
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • Risk area:
  • Rain (waterproof)
  • Rain (waterproof)
  • Rain (waterproof)
  • Rain (waterproof)
  • Rain (waterproof)
  • Rain (waterproof)
  • Rain (waterproof)
  • Rain (waterproof)
  • Rain (waterproof)
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Chest long
  • Chest long
  • Chest long
  • Chest long
  • Chest long
  • Chest long
  • Chest long
  • Chest long
  • Chest long
  • Fabric:
  • Teroflex
  • Teroflex
  • Teroflex
  • Teroflex
  • Teroflex
  • Teroflex
  • Teroflex
  • Teroflex
  • Teroflex
  • Weight:
  • 700 g/m²
  • 700 g/m²
  • 700 g/m²
  • 700 g/m²
  • 700 g/m²
  • 700 g/m²
  • 700 g/m²
  • 700 g/m²
  • 700 g/m²

 

Frequently Bought Together