ContactUsLinkComponent
Blotting papers, grade 556, Whatman™
Blotting papers, grade 556, Whatman™
Katalognr. 515-8110
undefined
Blotting papers, grade 556, Whatman™
Katalognr. 515-8110
Specifications
  • Description:
    Grade 556
  • Dimensions:
    37×100 mm
  • Seq:
    01
  • Pk:
    100
Specifications
About this item
Absorbent paper strips made from cellulose for the removal of excess liquids, e.g., from microscope specimens.