Specifications
- Pk:96
- Primary antibody reactivity:Mouse
- Target protein:IL-17F
- Description:Mouse IL-17F ELISA Kit
- Detection range:31,2 - 2000 pg/ml
- Sensitivity:<10 pg/ml
Specifications
About this item
Mouse IL-17F ELISA kit for the quantitative detection of mouse IL-17F in cell culture supernates, serum and plasma (heparin, EDTA).
Interleukin 17F, also called IL17F is involved in the regulation of normal versus aberrant T-cell responses. This gene is mapped to 6p12.2. The protein encoded by this gene is a cytokine that shares sequence similarity with IL17. This cytokine is expressed by activated T cells, and has been shown to stimulate the production of several other cytokines, including IL6, IL8, and CSF2/GM_CSF. This cytokine is also found to inhibit the angiogenesis of endothelial cells and induce endothelial cells to produce IL2, TGFB1/TGFB, and monocyte chemoattractant protein-1. It is suggested that targeting IL17 and IL17F or antagonizing IL17R might mitigate neutrophil-mediated inflammation in CF.