ContactUsLinkComponent
Human CRP (C Reactive Protein) ELISA kit
Human CRP (C Reactive Protein) ELISA kit
  BIRBORB78731-96
 :  Biorbyt
 :  
Human CRP (C Reactive Protein) ELISA kit
  BIRBORB78731-96
 :  Biorbyt
 :  
  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human CRP (C Reactive Protein) ELISA Kit
  • Detection range:
    2 - 600 pg/ml
  • Sensitivity:
    2 pg/ml
Human CRP (C Reactive Protein) ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CRP in serum, plasma, cell culture supernatants and urine.
 : Kit includes precoated 96-well microplate, wash buffer concentrate 20X, 25 ml; standards, 2 vials; assay diluent D 5X, 2×15 ml; assay diluent B 5X, 15 ml; detection antibody CRP, 2 vials; HRP-streptavidin concentrate 300X, 200 µl; TMB one-step substrate reagent, 12 ml; and stop solution, 8 ml.