ContactUsLinkComponent
Human alpha 1 Fetoprotein ELISA kit
Catalog # BIRBORB78656-96
Anbieter:  Biorbyt
CAS-Nummer:  
Human alpha 1 Fetoprotein ELISA kit
Catalog # BIRBORB78656-96
Anbieter:  Biorbyt
CAS-Nummer:  
Technische Daten
  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human Alpha 1 Fetoprotein ELISA Kit
Technische Daten
Über diesen Artikel
Human Alpha 1 Fetoprotein (AFP) ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of AFP in biological samples such as serum, plasma or cell culture supernatants.
Delivery: Kit includes pre-coated 96-well strip microplate, Wash Buffer Concentrate 20X, 25 ml; standards, 2 vials; Assay Diluent A, 30 ml; Assay Diluent B 5X, 15 ml; detection antibody, 2 vials; HRP-streptavidin concentrate, 200 µl; TMB One-Step Substrate Reagent, 12 ml; and Stop Solution, 8 ml.