Order Entry
Switzerland
ContactUsLinkComponent
Human FGF-21 ELISA kit
Human FGF-21 ELISA kit
Catalog # BIRBORB138095-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human FGF-21 ELISA kit
Catalog # BIRBORB138095-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human FGF-21 ELISA Kit
  • Detection range:
    31,2 - 2000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human FGF-21 ELISA Kit for the quantitative detection of human Fibroblast growth factor 21 (FGF-21) in cell culture supernates, serum and plasma (heparin, EDTA).

FGF-21 is a protein that in humans is encoded by the FGF21 gene. By genomic sequence analysis, Nishimura et al. (2000) identified the FGF21 gene within the 5-prime flanking region of the FUT1 gene on chromosome 19. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumour growth and invasion.

FGF21 stimulates glucose uptake in adipocytes but not in other cell types. This effect is additive to the activity of insulin. FGF21 treatment of adipocytes is associated with phosphorylation of FRS2, a protein linking FGF receptors to the Ras/MAP kinase pathway. FGF21 injection in ob/ob mice results in an increase in Glut1 in adipose tissue. FGF21 also protects animals from diet-induced obesity when overexpressed in transgenic mice and lowers blood glucose and triglyceride levels when administered to diabetic rodents.