ContactUsLinkComponent
VWR® Rota®, pH indicator paper, 1 to 14
VWR® Rota®, pH indicator paper, 1 to 14
Catalog # 85402.600
Supplier:  VWR Chemicals
undefined
VWR® Rota®, pH indicator paper, 1 to 14
Catalog # 85402.600
Supplier:  VWR Chemicals
Specifications
  • pH gradation:
    1 · 2 · 3 · 5 · 6 · 7 · 8 · 9 · 10 · 12 · 14
  • pH range:
    1 - 14
  • Pk:
    5
Specifications
About this item
Indicator paper for testing across the complete pH range.
  • Easy to use
  • Produced from high quality paper to give good colour matching of paper to printed calibration scale
Delivery: Supplied in plastic reels that ensure protection and long term stability. Reel of 5 m length, width: 7 mm.