ContactUsLinkComponent
Taq DNA polymerase, Cheetah™ HotStart
Taq DNA polymerase, Cheetah™ HotStart
Catalog # 29050.
Supplier:  Biotium
CAS Number:  
Taq DNA polymerase, Cheetah™ HotStart
Catalog # 29050.
Supplier:  Biotium
CAS Number:  
Specifications
  • Pk:
    500 EU
  • Description:
    Cheetah™ HotStart Taq DNA polymerase
Specifications
About this item
Cheetah™ Taq is a chemically modified hot-start Taq DNA polymerase useful for preventing or reducing nonspecific DNA amplification in PCR. Modified using a novel modifying reagent, Cheetah™ Taq represents a major improvement upon AmpliTaq Gold by having a faster activation time and better shelf-life.
Cheetah™ Taq regains both the 3′-5′ polymerase and 5′-exonuclease activities within two minutes. Cheetah™ Taq is also superior to antibody-based hot-start DNA polymerases due to its complete supression of enzyme activity at room temperature and full recovery of enzyme activity following activation.
Cheetah™ Taq is a patent-pending invention completely independent of AmpliTaq® Gold and related technologies.