Human NCK2 (from E. coli)
Catalog # BIRBORB80834-5
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:5 µG
- Source:E. coli
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein synonyms:Cytoplasmic
- Protein/peptide name:NCK2
- Formulation:NCK2 protein at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
- Tested applications:SDS-PAGE