Human adiponectin (Globular) (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:For long term, store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time two weeks.
- Biological activity:gAcrp30 activity is determined by its ability to inhibit the proliferation of mouse M1 cells. The expected ED50 for this effect is 1.0 2.5 ug/ml.
- Gene ID:9370
- Protein synonyms:ACDC
- Protein/peptide name:Adiponectin (globular)
- Purity:Greater than 97% as determined by HPLC and SDS PAGE.
- Solubility:We recommended reconstituting gAcrp30 at concentration of 0.1-1mg per ml with 5mM Tris pH-8 and 0.75mM DTT which can then be further diluted to other aqueous solutions.
- Sequence:MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN.
- Formulation:Sterile filtered and lyophilized from 1mg/ml in 10mM Tris pH-8 0.75mM DTT.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together