Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant AITRL (from E. coli)
Human recombinant AITRL (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant AITRL (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized AITRL although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution AITRL should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Protein synonyms:
    Osteostat
  • Protein/peptide name:
    AITRL
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    Add 0.2 uml of 0.1M Acetate buffer pH-4 and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to concentration of 10ug/ml. In higher concentrations the solubility of thi
  • Sequence:
    MRGSHHHHHH GMASTAKEPC MAKFGPLPSK WQMASSEPPC VNKVSDWKLE ILQNGLYLIY GQVAPNANYNDVAPFEVRLY KNKDMIQTLT NKSKIQNVGG TYELHVGDTI DLIFNSEHQV LKNNTYWGII LLANPQFIS.
  • Formulation:
    Lyophilized from 0.5 mg/ml in 0.03M Acetate buffer, pH-4.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together