Order Entry
Switzerland
ContactUsLinkComponent
Human soluble IL6 receptor alpha (from E. coli)
Human soluble IL6 receptor alpha (from E. coli)
Supplier:  Biorbyt
Certificates
Human soluble IL6 receptor alpha (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
  • Protein synonyms:
    Interleukin BSF 2
  • Protein/peptide name:
    Soluble IL6 Receptor alpha
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Formulation:
    CD126 His-Tag protein is supplied in PBS 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together