Human PF 4 (from platelets)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Platelets
- Species:Human
- Storage conditions:Human CXCL4 although stable at 25C week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
- Biological activity:Binding of HIT-antibodies.
- Protein synonyms:CXCL4
- Protein/peptide name:PF 4
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized CXCL4 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first four N-terminal amino acids was determined and was found to be Glu-Ala-Glu-Glu.
- Formulation:The CXCL4 protein was lyophilized in PBS buffer pH-7.4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together