Sheep leptin (from E. coli)
Marketsource
Supplier: Biorbyt
About this item
2 Options Available Below
Product Details & Documents
Documents
orb80492
Scheda dati
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- BIRBORB80492-100
- BIRBORB80492-1
- Protein/peptide name:
- Leptin
- Leptin
- Protein synonyms:
- LEP
- LEP
- Species:
- Sheep
- Sheep
- Source:
- E. coli
- E. coli
- Sequence:
- The sequence of the first five N-terminal amino acids was determined and was found to be Ala-Val-Pro-Ile-Arg.
- The sequence of the first five N-terminal amino acids was determined and was found to be Ala-Val-Pro-Ile-Arg.
- Biological activity:
- Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
- Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
- Purity:
- Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Formulation:
- The protein was lyophilized from concentrated (1mg/ml) solution with 0.0045mM NaHCO3.
- The protein was lyophilized from concentrated (1mg/ml) solution with 0.0045mM NaHCO3.
- Storage conditions:
- Lyophilized Leptin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Leptin should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Lyophilized Leptin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Leptin should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Solubility:
- It is recommended to reconstitute the lyophilized Leptin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- It is recommended to reconstitute the lyophilized Leptin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Tested applications:
- SDS-PAGE
- SDS-PAGE
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.BIRBORB80492-100Protein/peptide nameLeptinProtein synonymsLEPSpeciesSheepSourceE. coliSequenceThe sequence of the first five N-terminal amino acids was determined and was found to be Ala-Val-Pro-Ile-Arg.Biological activityBiological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.PurityGreater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGEFormulationThe protein was lyophilized from concentrated (1mg/ml) solution with 0.0045mM NaHCO3.Storage conditionsLyophilized Leptin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Leptin should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.SolubilityIt is recommended to reconstitute the lyophilized Leptin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.Tested applicationsSDS-PAGEMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.BIRBORB80492-1Protein/peptide nameLeptinProtein synonymsLEPSpeciesSheepSourceE. coliSequenceThe sequence of the first five N-terminal amino acids was determined and was found to be Ala-Val-Pro-Ile-Arg.Biological activityBiological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.PurityGreater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGEFormulationThe protein was lyophilized from concentrated (1mg/ml) solution with 0.0045mM NaHCO3.Storage conditionsLyophilized Leptin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Leptin should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.SolubilityIt is recommended to reconstitute the lyophilized Leptin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.Tested applicationsSDS-PAGEMarketsource



