Rat recombinant Cystatin C (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/ thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. Minimum shelf life years.
- Protein synonyms:AD8
- Protein/peptide name:Cystatin C
- Purity:Greater than 96% as determined by SDS-PAGE.
- Solubility:Add 0.2 uml of 0.1M Acetate buffer pH4 and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to concentration of 0.1mg/ml. In higher concentrations the solubility of thi
- Sequence:MRGSHHHHHH GMASGTSRPP PRLLGAPQEA DASEEGVQRA LDFAVSEYNK GSNDAYHSRA IQVVRARKQLVAGINYYLDV EMGRTTCTKS QTNLTNCPFH DQPHLMRKAL CSFQIYSVPW KGTHTLTKSS.
- Formulation:Filtered (0.4 umicron) and lyophilized from 0.5 mg/ml in 0.03M Acetate buffer, pH 4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together