Human Intrinsic Factor (from Insect cells (Sf9))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Insect cells (Sf9)
- Species:Human
- Storage conditions:IF although stable at 10 °C for week, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biological activity:Standard ELISA test (checker-board analysis of positive/negative sera panels), immunodot with anti-IF autoantibody positive patient sera.Can be used for solid ELISA and fluid phase diagnostic assays, 0.25-0.65ug/ml.Suitable for biotinylation and iodination.
- Protein synonyms:Gastric intrinsic factor
- Protein/peptide name:Intrinsic Factor
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Formulation:The protein solution (0.7mg/ml) contains 20 mM HEPES pH-8, 100mM NaCl, 20% Glycerol.
- Tested applications:ELISA , SDS-PAGE
Specifications
Frequently Bought Together