Human recombinant PKC-e (from insect cells (Sf9))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein synonyms:Protein kinase epsilon type
- Protein/peptide name:PKC-e
- Purity:Greater than 90% as determined by SDS-PAGE.
- Formulation:PRKCE is supplied in 20 mM Tris, pH 7.4, 0.1M NaCl, 20% glycerol, 1mM DTT, 0.1mM EDTA, 0.2 um PMSF, 1mM benzamidine and 0.03% Brij-35.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together