Human TGF beta receptor 2 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized TGFBR2 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution TGFBR2 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biological activity:In ELISA assay, concentrations 0.5-1ug/ml can be used for TGF-beta measurements
- Protein synonyms:AAT3
- Protein/peptide name:TGF beta Receptor 2
- Purity:Greater than 97.0% as determined by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized TGFBR2 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:HVQK SVNNDMIVTD NNGAVKFPQL CKFCDVRFST CDNQKSCMSN CSITSICEKP QEVCVAVWRK NDENITLETV CHDPKLPYHD FILEDAASPK CIMKEKKKPG ETFFMCSCSS DECNDNIIFS EEYNTSNPD
- Formulation:Lyophilized from 0.2 um filtered solution in buffer containing 150mM NaCl 50mM sodium phosphate buffer pH-7.5.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together