Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant CTGF HEK (from HEK293 cells)
Human recombinant CTGF HEK (from HEK293 cells)
Supplier:  Biorbyt
Certificates
Human recombinant CTGF HEK (from HEK293 cells)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized CTGF remains stable until the expiry date when stored at −20 °C.
  • Protein synonyms:
    CCN2
  • Protein/peptide name:
    CTGF HEK
  • Purity:
    Greater than 90% as determined by SDS-PAGE.
  • Solubility:
    The CTGF can be solubilized with sterile pyrogen free water at concentration of 0.5mg/ml, let pellet dissolve completely. Protein is not sterile! Please filter the product by an appropriate sterile filter before using it in the cell culture.
  • Sequence:
    QNCSGPCRCPDEPAP RCPAGVSLVLDGCGCCR VCAKQLGELCTER DPCDPHKGLFCDFGSPAN RKIGVCTAKDGAP CIFGGTVYRSGES FQSSCKYQCTCLDGAV GCMPLCSMDVRLPSPDCPFPRRVK LPGKCCEEWVCDEP KDQTVVGPALAAYR LEDTFGPDPTMIRAN CLVQTTEWSACSKTCGMGIS TRVTNDNASCRLEK QSRLCMVRPCEAD LEENIKKGKKCIR TPKISKPIKFELSGCTSMKTY RAKFCGVCTDGRCCT PHRTTTLPVEFKCPDGEV MKKNMMFIKTCAC HYNCPGDNDIFES LYYRKMYGDMAAA ADYKDDDDK.
  • Formulation:
    Filtered and lyophilized from 0.5mg/ml in 20 mM Tris buffer and 50mM NaCl, pH-7.5.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together