Human IL32 alpha (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized IL32 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution IL32 should be stored at 4 °C between 2-7 days and for future use below −18 °C. Please prevent freeze/thaw cycles.
- Biological activity:Human IL-32 alpha activity is measured via the dose-dependent induction of TNF-alpha in the human THP-1 monocytic cell line.
- Protein synonyms:NK4
- Protein/peptide name:IL32 alpha
- Purity:Greater than 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized IL-32 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MCFPKVLSDD MKKLKARMHQ AIERFYDKMQ NAESGRGQVM SSLAELEDDF KEGYLETVAA YYEEQHPELT PLLEKERDGL RCRGNRSPVP DVEDPATEEP GESFCDKSYG APRGDKEELT PQKCSEPQSS K.
- Formulation:IL-32 was lyophilized from concentrated (1mg/ml) solution in water containing 50mM sodium Phosphate buffer pH=7.5.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together