Rat MCP1 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Rat
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Biological activity:ED50 1.0-10.0 ng/mL. The biological activity was determined by measuring the dose dependent chemotaxis with human THP-1 cells. The optimal concentration should be determined for each specific application by an initial dose-response assay.
- Protein synonyms:C-C motif chemokine 2
- Protein/peptide name:MCP1
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized Monocyte Chemotactic Protein-1 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be Gln-Pro-Asp-Ala-Val.
- Formulation:The protein was lyophilized from concentrated (1mg/ml) sterile solution containing no additives.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together