Order Entry
Switzerland
ContactUsLinkComponent
 
HPV 16 (from E. coli)
 :  Biorbyt
 
HPV 16 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Storage conditions:
    Two years when when kept frozen at -2 °C to −80 °C.
  • Protein synonyms:
    Papillomavirus
  • Protein/peptide name:
    HPV 16
  • Purity:
    HPV16 protein purity is greater than 95% pure as determined by 10% PAGE (Coomassie staining). The recombinant HPV-16 fusion protein was purified by GSH affinity chromatography technique.
  • Sequence:
    MQVTFIYILVITCYENDVNVYHIFFQMSLWLPSEATVYLPPVPVSKVVSTDEYVARTNIYYHAGTSRLLAVGHPYFPIKKPNNNKILVPKVSGLQYRVFRIHLPDPNKFGFPDTSFYNPDTQRLVWACVGVEVGRGQPLGVGISGHPLLNKLDDTENASAYAANAGVDNRECISMDYKQTQLCLIGCKPPIGEHWGKGSPCTNVAVNPGDCPPLELINTVIQDGDMVHTGFGAMDFTTLQANKSEVPLDICTSICKYPDYIKMVSEPYGDSLFFYLRREQMFVRHLFNRAGTVGENVPDDLYIKGSGSTANLASSNYFPTPSGSMVTSDAQIFNKPYWLQRAQGHNNGICWGNQLFVTVVDTTRSTNMSLCAAISTSETTYKNTNFKEYLRHGEEYDLQFIFQLCKITLTADVMTYIHSMNSTILEDWNFGLQPPPGGTLEDTYRFVTQAIACQKHTPPAPKEDDPLKKYTFWEVNLKEKFSADLDQFPLGRKFLLQAGLKAKPKFTLGKRKATPTTSSTSTTAKRKKRKL
  • Formulation:
    The protein is formulated with 1x PBS and 0.5mM EDTA.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together