HIV-2 Envelope (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Storage conditions:HIV-2 Envelope protein is shipped at ambient temperature. Upon arrival, Store at −20 °C Stable for Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HIV-2 Envelope
- Purity:Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
- Formulation:50mM Tris-HCL 6M urea, pH-7.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together