HIV-1 Integrase (from E. coli)
: Biorbyt
- Source:E. coli
- Storage conditions:Protein is shipped at ambient temperature. Upon arrival, Store at −20 °C Stable for Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HIV-1 Integrase
- Purity:Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
- Formulation:1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X 50% Glycerol.
- Tested applications:SDS-PAGE
Frequently Bought Together