Order Entry
Switzerland
ContactUsLinkComponent
HAV P2C-P3A
HAV P2C-P3A
Supplier:  Biorbyt
Certificates
HAV P2C-P3A
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    HAV P2C-P3A
  • Purity:
    HAV P2C-P3A protein is >90% pure as determined by 10% PAGE (coomassie staining). HAV P2C-P3A protein was purified by proprietary chromatographic technique.
  • Formulation:
    10mM CBB, pH9.6, 0.1% SDS and 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together