Dengue envelope-3 22 kDa
Supplier: Biorbyt
Certificates
About this item
Specifications
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:Dengue Envelope-3 22kDa
- Purity:Protein is >95% pure as determined by 10% PAGE (coomassie staining).
- Sequence:GSLATLRKLC IEGKITNITT DSRCPTQGEA VLPEEQDQNY VCKHTYVDRG WGNGCGLFGK GSLVTCAKFQ CLEPIEGKVV QYENLKYTVI ITVHTGDQHQ VGNETQGVTA EITPQASTTE AILPEYGTLG LECSPRTGLD FNEMILLTMK NKAWMVHRQW FFDLPLPWTS GATTETPTWN RKELLVTFKN AHAKKQE.
- Formulation:50mM sodium chloride-phosphorous buffer, pH-7.5
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together