Order Entry
Switzerland
ContactUsLinkComponent
HIV-1 p17/ p24/ gp120
HIV-1 p17/ p24/ gp120
  BIRBORB85297-100
 :  Biorbyt
 :  
HIV-1 p17/ p24/ gp120
  BIRBORB85297-100
 :  Biorbyt
 :  

 

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Storage conditions:
    Store at−20 °C.
  • Protein synonyms:
    Immunodeficiency Virus 1 antigen
  • Protein/peptide name:
    HIV-1 p17/ p24/ gp120
  • Purity:
    Greater than 95% by SDS-PAGE, and RP-HPLC.
  • Formulation:
    Liquid. Supplied in 20 mM PBS, pH 7.8, 0.5 M NaCl,1 mM DTT, and 8 M urea.
  • Tested applications:
    WB , SDS-PAGE