Order Entry
Switzerland
ContactUsLinkComponent
Recombinant HIV-1 p24,Biotin
Recombinant HIV-1 p24,Biotin
Catalog # BIRBORB85317-100
Supplier:  Biorbyt
CAS Number:  
Recombinant HIV-1 p24,Biotin
Catalog # BIRBORB85317-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at−20 °C.
  • Protein synonyms:
    Immunodeficiency Virus 1 antigen
  • Protein/peptide name:
    HIV-1 p24
  • Purity:
    Greater than 95% by SDS-PAGE and RP-HPLC
  • Formulation:
    Liquid. Supplied in 50 mM Tris-HCl pH 7.2 and 8 Murea.
  • Tested applications:
    WB , SDS-PAGE

Specifications

About this item