Drosophila melanogaster Recombinant GAGA-POZ (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Drosophila melanogaster
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Avoid multiple freeze-thaw cycles.
- Protein synonyms:Transcription factor GAGA
- Protein/peptide name:GAGA-POZ
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Sequence:MSLPMNSLYS LTWGDYGTSL VSAIQLLRCH GDLVDCTLAA GGRSFPAHKI VLCAASPFLLDLLKNTPCKH PVVMLAGVNA NDLEALLEFV YRGEVSVDHA QLPSLLQAAQ CLNIQGLAPQTVTKDDYTTH.
- Formulation:The protein containing 10mM HEPES (pH-7.4) and 25mM NaCl.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together