Human HDL (from plasma)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Plasma
- Species:Human
- Storage conditions:Human HDL although stable at 4 °C for week, should be stored desiccated below -15 °C. (short term i. e. 3 months) and below −80 °C for long term.
- Protein synonyms:High Density Lipoprotein
- Protein/peptide name:HDL
- Purity:Cholesterol level >200mg/l. <1% LDL.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together