Human EGF receptor-Sf9 (ErbB1) (from Baculovirus (Sf9 Insect cells))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Baculovirus (Sf9 Insect cells)
- Species:Human
- Storage conditions:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year. Please avoid freeze-thaw cycles.
- Biological activity:Determination of Km value by Filter binding assay MAFC membrane.
- Protein synonyms:Epidermal growth factor receptor
- Protein/peptide name:EGF Receptor-Sf9 (ErbB1)
- Formulation:EGFR in 50mM Tris-HCl, pH 8.0, 100mM NaCl, 5mM DTT, 15mM reduced glutathione and 20% glycerol.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together