Human recombinant E7
Katalognr. BIRBORB244751-50
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Storage conditions:Store at −20 °C. for extended storage, conserve at −20 °C or −80 °C.
- UniProtKB:P06788
- Protein/peptide name:E7
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:16.1 kDa
- Sequence:MHGPKATLQDIVLHLEPQNEIPVDLLCHEQLSDSEEENDEIDGVNHQHLPARRAEPQRHTMLCMCCKCEARIKLVVESSADDLRAFQQLFLNTLSFVCPWCASQQ
- Formulation:20mM Tris-HCl based buffer, pH8.0