Rat SCF (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Gene ID:60427
- Protein synonyms:Stem Cell Factor
- Protein/peptide name:SCF
- Purity:> 95% by SDS-PAGE and HPLC analyses
- Molecular weight:18.5 kDa
- Sequence:MQEICRNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMVTHLSVSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVAC MEENAPKNVK ESLKKPETRNFTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4
- Tested applications:SDS-PAGE , HPLC
Frequently Bought Together