Rhesus macaque Rhesus IL4 (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Rhesus monkey
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:B cell growth factor 1
- Protein/peptide name:Rhesus IL4
- Purity:> 96 % by SDS-PAGE and HPLC analyses
- Molecular weight:15.0 kDa
- Sequence:HNCHIALREI IETLNSLTEQ KTLCTKLTIT DILAASKNTT EKETFCRAAT VLRQFYSHHEKDTRCLGATA QQFHRHKQLI RFLKRLDRNL WGLAGLNSCP VKEANQSTLE DFLERLKTIMREKYSKCSS
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, 3 % trehalose
- Tested applications:SDS-PAGE , HPLC
Frequently Bought Together