Rat BD4 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:beta-Defensin 4
- Protein/peptide name:BD4
- Purity:> 95% by SDS-PAGE and HPLC analyses
- Molecular weight:4.4KDa
- Sequence:QSINNPITCL TKGGVCWGPC TGGFRQIGTC GLPRVRCCKK K
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in 10mM PB, pH 7.4, 500mM NaCl.
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together