Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Artificial troponin I-T-C complex
Artificial troponin I-T-C complex
Artificial troponin I-T-C complex
BIRBORB79700-0.05
:
Biorbyt
:
Artificial troponin I-T-C complex
BIRBORB79700-0.05
:
Biorbyt
:
Pk:
0,5 mg
Storage conditions:
Store at -2 to -8 °C.
Protein/peptide name:
Artificial troponin I-T-C complex
Purity:
Greater than 95% pure SDS-PAGE. Protein G Chromatography